Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGCCGGCAAACCTTTTCCTTCAC[C/T]TCCTCAGCCTCACTGGCCAATTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604528 MIM: 604037 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNH4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNH4 - potassium voltage-gated channel subfamily H member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012285.2 | 2958 | Silent Mutation | GAA,GAG | E,E 875 | NP_036417.1 | |
XM_017024400.1 | 2958 | Silent Mutation | GAA,GAG | E,E 875 | XP_016879889.1 | |
XM_017024401.1 | 2958 | Silent Mutation | GAA,GAG | E,E 826 | XP_016879890.1 | |
XM_017024402.1 | 2958 | Silent Mutation | GAA,GAG | E,E 815 | XP_016879891.1 | |
XM_017024403.1 | 2958 | Intron | XP_016879892.1 |
RAB5C - RAB5C, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |