Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCGTGGCAGTATCGCTTCCCACC[C/G]TTCTTTACGTGAGGCTCAGACCCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610907 MIM: 601844 | ||||||||||||||||||||
Literature Links: |
VPS25 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
VPS25 - vacuolar protein sorting 25 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032353.3 | 51 | Silent Mutation | CCC,CCG | P,P 15 | NP_115729.1 | |
XM_017025224.1 | 51 | Silent Mutation | CCC,CCG | P,P 15 | XP_016880713.1 |
WNK4 - WNK lysine deficient protein kinase 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |