Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGTAGAGGTTTATTTCAACAAAA[G/T]GTGTAAAGAGTGCATCTGAAGCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 200350 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACACA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACACA - acetyl-CoA carboxylase alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198834.2 | 10628 | UTR 3 | NP_942131.1 | |||
NM_198836.2 | 10628 | UTR 3 | NP_942133.1 | |||
NM_198837.1 | 10628 | UTR 3 | NP_942134.1 | |||
NM_198838.1 | 10628 | UTR 3 | NP_942135.1 | |||
NM_198839.2 | 10628 | UTR 3 | NP_942136.1 | |||
XM_005257267.4 | 10628 | UTR 3 | XP_005257324.1 | |||
XM_006721853.1 | 10628 | UTR 3 | XP_006721916.1 | |||
XM_011524701.1 | 10628 | UTR 3 | XP_011523003.1 | |||
XM_011524703.1 | 10628 | UTR 3 | XP_011523005.1 | |||
XM_011524704.2 | 10628 | UTR 3 | XP_011523006.1 | |||
XM_017024553.1 | 10628 | UTR 3 | XP_016880042.1 | |||
XM_017024554.1 | 10628 | UTR 3 | XP_016880043.1 | |||
XM_017024555.1 | 10628 | UTR 3 | XP_016880044.1 |