Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTAGGACCAAGGAAAAGCTTCC[C/T]AAGGGGCTCCCAGTGCTGGCTGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611045 MIM: 605315 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
G6PC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
G6PC3 - glucose-6-phosphatase catalytic subunit 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HDAC5 - histone deacetylase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001015053.1 | Intron | NP_001015053.1 | ||||
NM_005474.4 | Intron | NP_005465.2 | ||||
XM_005256905.1 | Intron | XP_005256962.1 | ||||
XM_005256906.4 | Intron | XP_005256963.1 | ||||
XM_011524149.1 | Intron | XP_011522451.1 | ||||
XM_011524150.1 | Intron | XP_011522452.1 | ||||
XM_017023988.1 | Intron | XP_016879477.1 | ||||
XM_017023989.1 | Intron | XP_016879478.1 | ||||
XM_017023990.1 | Intron | XP_016879479.1 | ||||
XM_017023991.1 | Intron | XP_016879480.1 | ||||
XM_017023992.1 | Intron | XP_016879481.1 | ||||
XM_017023993.1 | Intron | XP_016879482.1 | ||||
XM_017023994.1 | Intron | XP_016879483.1 | ||||
XM_017023995.1 | Intron | XP_016879484.1 | ||||
XM_017023996.1 | Intron | XP_016879485.1 |