Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTAGGCAGCCTTCCCTTTGCCCC[C/T]TTTCTTCCCTTCCAGGATTAGTGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C17orf53 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C17orf53 - chromosome 17 open reading frame 53 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171251.2 | Intron | NP_001164722.1 | ||||
NM_001321310.1 | Intron | NP_001308239.1 | ||||
NM_001321311.1 | Intron | NP_001308240.1 | ||||
NM_024032.4 | Intron | NP_076937.2 | ||||
XM_011525187.1 | Intron | XP_011523489.1 | ||||
XM_011525188.1 | Intron | XP_011523490.1 | ||||
XM_011525189.1 | Intron | XP_011523491.1 | ||||
XM_011525190.1 | Intron | XP_011523492.1 | ||||
XM_011525191.2 | Intron | XP_011523493.1 | ||||
XM_011525192.1 | Intron | XP_011523494.1 | ||||
XM_011525193.1 | Intron | XP_011523495.1 | ||||
XM_011525194.1 | Intron | XP_011523496.1 | ||||
XM_011525195.1 | Intron | XP_011523497.1 | ||||
XM_011525197.2 | Intron | XP_011523499.1 | ||||
XM_017025031.1 | Intron | XP_016880520.1 | ||||
XM_017025032.1 | Intron | XP_016880521.1 |