Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAGAGAGTGTGTATTAGGATCCC[A/C]TCTAGTGGCTTTGGTACCAAGGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614677 MIM: 603892 MIM: 137780 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC103 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC103 - coiled-coil domain containing 103 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EFTUD2 - elongation factor Tu GTP binding domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GFAP - glial fibrillary acidic protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001131019.2 | Intron | NP_001124491.1 | ||||
NM_001242376.1 | Intron | NP_001229305.1 | ||||
NM_002055.4 | Intron | NP_002046.1 | ||||
XM_017024451.1 | Intron | XP_016879940.1 |