Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATTTAAATGTTAAATGTTAAGTG[A/C]AAAACTTTAAAATTTTTAAAAGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614956 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLFN12L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLFN12L - schlafen family member 12 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195790.1 | Intron | NP_001182719.1 | ||||
XM_017024005.1 | Intron | XP_016879494.1 | ||||
XM_017024006.1 | Intron | XP_016879495.1 | ||||
XM_017024007.1 | Intron | XP_016879496.1 | ||||
XM_017024008.1 | Intron | XP_016879497.1 | ||||
XM_017024009.1 | Intron | XP_016879498.1 |