Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGATTAAATTCTGCTCAGTAGTT[A/G]CAAGTAACTGAACATTTTTTTCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607427 MIM: 164880 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENOSF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ENOSF1 - enolase superfamily member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
YES1 - YES proto-oncogene 1, Src family tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005433.3 | 4632 | UTR 3 | NP_005424.1 | |||
XM_005258139.3 | 4632 | UTR 3 | XP_005258196.1 | |||
XM_017025960.1 | 4632 | UTR 3 | XP_016881449.1 |