Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGCCAGTTATAAATAAATAAAT[C/T]ACATCATTTATCTTAGAGCAGAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194534 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF24 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF24 - zinc finger protein 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308123.1 | 3305 | Intron | NP_001295052.1 | |||
NM_006965.3 | 3305 | UTR 3 | NP_008896.2 | |||
XM_005258341.3 | 3305 | UTR 3 | XP_005258398.1 |