Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGTTCAGGTGTGAGAGTTTCCTT[C/T]GTGCCTATCTTGGTGGGTGAGCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609194 MIM: 615430 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CABLES1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CABLES1 - Cdk5 and Abl enzyme substrate 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100619.2 | Intron | NP_001094089.1 | ||||
NM_001256438.1 | Intron | NP_001243367.1 | ||||
NM_138375.2 | Intron | NP_612384.1 |
TMEM241 - transmembrane protein 241 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318834.1 | Intron | NP_001305763.1 | ||||
NM_032933.5 | Intron | NP_116322.3 | ||||
XM_011526233.2 | Intron | XP_011524535.1 | ||||
XM_017026043.1 | Intron | XP_016881532.1 | ||||
XM_017026044.1 | Intron | XP_016881533.1 | ||||
XM_017026045.1 | Intron | XP_016881534.1 | ||||
XM_017026046.1 | Intron | XP_016881535.1 | ||||
XM_017026047.1 | Intron | XP_016881536.1 | ||||
XM_017026048.1 | Intron | XP_016881537.1 | ||||
XM_017026049.1 | Intron | XP_016881538.1 | ||||
XM_017026050.1 | Intron | XP_016881539.1 | ||||
XM_017026051.1 | Intron | XP_016881540.1 |