Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTCCTGGTCCACGTCGGCTATCT[C/G]GTGCTTCCAGTGTTTGGCTCTGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF165 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNF165 - ring finger protein 165 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256758.1 | 78 | UTR 5 | NP_001243687.1 | |||
NM_152470.2 | 78 | Silent Mutation | CTC,CTG | L,L 9 | NP_689683.2 | |
XM_011526016.2 | 78 | Silent Mutation | CTC,CTG | L,L 9 | XP_011524318.1 | |
XM_017025788.1 | 78 | Silent Mutation | CTC,CTG | L,L 9 | XP_016881277.1 |