Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGCTGCCCCGCTCGAAGAGGTTG[C/T]GCATTTTCTGCTTCAGCTCCCGCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613072 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOXHD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOXHD1 - lipoxygenase homology domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145472.2 | 3204 | Missense Mutation | CAC,CGC | H,R 1084 | NP_001138944.1 | |
NM_001145473.2 | 3204 | Missense Mutation | CAC,CGC | H,R 434 | NP_001138945.1 | |
NM_001173129.1 | 3204 | Missense Mutation | CAC,CGC | H,R 434 | NP_001166600.1 | |
NM_001308013.1 | 3204 | Missense Mutation | CAC,CGC | H,R 988 | NP_001294942.1 | |
NM_144612.6 | 3204 | Missense Mutation | CAC,CGC | H,R 2133 | NP_653213.6 | |
XM_006722388.3 | 3204 | Missense Mutation | CAC,CGC | H,R 1128 | XP_006722451.1 | |
XM_006722389.3 | 3204 | Missense Mutation | CAC,CGC | H,R 1084 | XP_006722452.1 | |
XM_006722390.3 | 3204 | Missense Mutation | CAC,CGC | H,R 1084 | XP_006722453.1 | |
XM_006722391.3 | 3204 | Missense Mutation | CAC,CGC | H,R 1066 | XP_006722454.1 | |
XM_011525804.2 | 3204 | Missense Mutation | CAC,CGC | H,R 1582 | XP_011524106.1 | |
XM_011525807.2 | 3204 | Missense Mutation | CAC,CGC | H,R 988 | XP_011524109.1 | |
XM_011525810.2 | 3204 | Missense Mutation | CAC,CGC | H,R 451 | XP_011524112.1 | |
XM_011525811.2 | 3204 | Missense Mutation | CAC,CGC | H,R 434 | XP_011524113.1 | |
XM_017025540.1 | 3204 | Missense Mutation | CAC,CGC | H,R 1847 | XP_016881029.1 | |
XM_017025541.1 | 3204 | Missense Mutation | CAC,CGC | H,R 1876 | XP_016881030.1 | |
XM_017025542.1 | 3204 | Intron | XP_016881031.1 | |||
XM_017025543.1 | 3204 | Intron | XP_016881032.1 | |||
XM_017025544.1 | 3204 | Intron | XP_016881033.1 | |||
XM_017025545.1 | 3204 | Intron | XP_016881034.1 | |||
XM_017025546.1 | 3204 | Intron | XP_016881035.1 | |||
XM_017025547.1 | 3204 | Intron | XP_016881036.1 | |||
XM_017025548.1 | 3204 | Missense Mutation | CAC,CGC | H,R 1989 | XP_016881037.1 |