Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGGGAAATGCAGTAGACGCCAGA[A/G]CATCAAAGGCCATGTATTTCCTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601366 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMAD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SMAD2 - SMAD family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003652.3 | 9714 | UTR 3 | NP_001003652.1 | |||
NM_001135937.2 | 9714 | UTR 3 | NP_001129409.1 | |||
NM_005901.5 | 9714 | UTR 3 | NP_005892.1 | |||
XM_005258259.3 | 9714 | UTR 3 | XP_005258316.1 | |||
XM_006722451.3 | 9714 | UTR 3 | XP_006722514.1 | |||
XM_011525984.2 | 9714 | UTR 3 | XP_011524286.1 | |||
XM_011525985.2 | 9714 | UTR 3 | XP_011524287.1 | |||
XM_017025745.1 | 9714 | UTR 3 | XP_016881234.1 | |||
XM_017025746.1 | 9714 | UTR 3 | XP_016881235.1 | |||
XM_017025747.1 | 9714 | UTR 3 | XP_016881236.1 | |||
XM_017025748.1 | 9714 | UTR 3 | XP_016881237.1 | |||
XM_017025749.1 | 9714 | Intron | XP_016881238.1 | |||
XM_017025750.1 | 9714 | UTR 3 | XP_016881239.1 |