Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCAAACATTTTTTAATTTTGACA[C/T]TAATTTCTGGCTTAATTCTACTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613903 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF540 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF540 - zinc finger protein 540 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172225.2 | Intron | NP_001165696.1 | ||||
NM_001172226.2 | Intron | NP_001165697.1 | ||||
NM_152606.4 | Intron | NP_689819.1 |
ZNF571-AS1 - ZNF571 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF793 - zinc finger protein 793 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |