Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTGTCTAAAATAAAGTATCTGTT[A/G]TTCTAAAGGGGCTACATGATAAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613903 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF540 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF540 - zinc finger protein 540 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172225.2 | Intron | NP_001165696.1 | ||||
NM_001172226.2 | Intron | NP_001165697.1 | ||||
NM_152606.4 | Intron | NP_689819.1 |
ZNF571 - zinc finger protein 571 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290314.1 | Intron | NP_001277243.1 | ||||
NM_001321272.1 | Intron | NP_001308201.1 | ||||
NM_016536.4 | Intron | NP_057620.3 | ||||
XM_011527024.2 | Intron | XP_011525326.1 | ||||
XM_017026855.1 | Intron | XP_016882344.1 | ||||
XM_017026856.1 | Intron | XP_016882345.1 | ||||
XM_017026857.1 | Intron | XP_016882346.1 | ||||
XM_017026858.1 | Intron | XP_016882347.1 | ||||
XM_017026859.1 | Intron | XP_016882348.1 | ||||
XM_017026860.1 | Intron | XP_016882349.1 |
ZNF571-AS1 - ZNF571 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |