Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGGTATAGTTACCTATTTGTAT[G/T]GATCTGTAAAAGCAAGATGGAACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF780A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF780A - zinc finger protein 780A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010880.2 | 6829 | Intron | NP_001010880.2 | |||
NM_001142577.1 | 6829 | Intron | NP_001136049.1 | |||
NM_001142578.1 | 6829 | Intron | NP_001136050.1 | |||
NM_001142579.1 | 6829 | UTR 3 | NP_001136051.1 | |||
XM_005258773.2 | 6829 | UTR 3 | XP_005258830.1 | |||
XM_006723150.3 | 6829 | UTR 3 | XP_006723213.1 | |||
XM_011526771.2 | 6829 | UTR 3 | XP_011525073.1 | |||
XM_011526772.2 | 6829 | UTR 3 | XP_011525074.1 | |||
XM_017026611.1 | 6829 | UTR 3 | XP_016882100.1 | |||
XM_017026612.1 | 6829 | UTR 3 | XP_016882101.1 | |||
XM_017026613.1 | 6829 | UTR 3 | XP_016882102.1 | |||
XM_017026614.1 | 6829 | UTR 3 | XP_016882103.1 | |||
XM_017026615.1 | 6829 | UTR 3 | XP_016882104.1 | |||
XM_017026616.1 | 6829 | UTR 3 | XP_016882105.1 | |||
XM_017026617.1 | 6829 | UTR 3 | XP_016882106.1 | |||
XM_017026618.1 | 6829 | UTR 3 | XP_016882107.1 | |||
XM_017026619.1 | 6829 | UTR 3 | XP_016882108.1 |