Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGCCAGGCAGCCCCACACTCCC[C/T]GCCAAGCCCTCCCCAAGCCCAGGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603074 | ||||||||||||||||||||
Literature Links: |
PAFAH1B3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PAFAH1B3 - platelet activating factor acetylhydrolase 1b catalytic subunit 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRR19 - proline rich 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199285.2 | 399 | Silent Mutation | CCC,CCT | P,P 103 | NP_954979.2 | |
XM_005258779.4 | 399 | Silent Mutation | CCC,CCT | P,P 155 | XP_005258836.2 | |
XM_005258780.3 | 399 | Silent Mutation | CCC,CCT | P,P 103 | XP_005258837.1 | |
XM_006723154.3 | 399 | Silent Mutation | CCC,CCT | P,P 103 | XP_006723217.1 | |
XM_011526787.1 | 399 | Silent Mutation | CCC,CCT | P,P 103 | XP_011525089.1 | |
XM_011526788.1 | 399 | Silent Mutation | CCC,CCT | P,P 103 | XP_011525090.1 | |
XM_011526789.2 | 399 | Silent Mutation | CCC,CCT | P,P 103 | XP_011525091.1 | |
XM_011526790.2 | 399 | Silent Mutation | CCC,CCT | P,P 103 | XP_011525092.1 |
TMEM145 - transmembrane protein 145 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |