Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACTTAACCCCTTTGCCTGAAGAT[G/T]CAAAATGTTGTTTTTAATCCCGTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF285 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF285 - zinc finger protein 285 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291488.1 | Intron | NP_001278417.1 | ||||
NM_001291489.1 | Intron | NP_001278418.1 | ||||
NM_001291490.1 | Intron | NP_001278419.1 | ||||
NM_001291491.1 | Intron | NP_001278420.1 | ||||
NM_152354.5 | Intron | NP_689567.4 |