Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGTGCCCTACATAAACCTCTGT[C/T]GAATGAATGAATAAATACCCTTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600551 MIM: 606580 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPR4 - G protein-coupled receptor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005282.2 | 3503 | UTR 3 | NP_005273.1 | |||
XM_017026607.1 | 3503 | UTR 3 | XP_016882096.1 | |||
XM_017026608.1 | 3503 | UTR 3 | XP_016882097.1 |
OPA3 - optic atrophy 3 (autosomal recessive, with chorea and spastic paraplegia) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |