Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGACCTTCGCAGACGTGGCTGTG[A/G]ACTTCACCAAAGAGGAGTGGACCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603996 | ||||||||||||||||||||
Literature Links: |
ZNF114 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF114 - zinc finger protein 114 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301062.1 | 389 | UTR 5 | NP_001287991.1 | |||
NM_153608.2 | 389 | Missense Mutation | AAC,GAC | N,D 14 | NP_705836.1 | |
XM_011526574.2 | 389 | Missense Mutation | AAC,GAC | N,D 60 | XP_011524876.1 | |
XM_017026414.1 | 389 | Missense Mutation | AAC,GAC | N,D 60 | XP_016881903.1 | |
XM_017026415.1 | 389 | Missense Mutation | AAC,GAC | N,D 14 | XP_016881904.1 |