Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGGCCACAGCGGTTGCAGGTCA[A/G]GCCTGTGACCCCTAACTTGCAGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
NTN5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NTN5 - netrin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145807.2 | 1115 | Silent Mutation | CTG,TTG | L,L 303 | NP_665806.1 | |
XM_011526443.2 | 1115 | Silent Mutation | CTG,TTG | L,L 303 | XP_011524745.1 | |
XM_017026274.1 | 1115 | Silent Mutation | CTG,TTG | L,L 148 | XP_016881763.1 |
SEC1P - secretory blood group 1, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |