Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCACATCTTCAAGTGTAGATGGTC[C/T]AGAGTGGCCATCTTCTCCCTGTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608253 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TSKS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TSKS - testis specific serine kinase substrate | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021733.1 | 1652 | Silent Mutation | CTA,CTG | L,L 547 | NP_068379.1 | |
XM_011527201.1 | 1652 | Silent Mutation | CTA,CTG | L,L 572 | XP_011525503.1 | |
XM_011527202.1 | 1652 | Missense Mutation | AGA,GGA | R,G 562 | XP_011525504.1 | |
XM_011527203.2 | 1652 | UTR 3 | XP_011525505.1 |