Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTATCAGAGGAGGAAGACTTACC[A/G]TTGGACAGCCCTGCCCTGGAGGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 160793 MIM: 174761 MIM: 606802 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYBPC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYBPC2 - myosin binding protein C, fast type | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLD1 - DAN polymerase delta 1, catalytic subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPIB - Spi-B transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243998.1 | 430 | Silent Mutation | CCA,CCG | P,P 44 | NP_001230927.1 | |
NM_001243999.1 | 430 | Silent Mutation | CCA,CCG | P,P 135 | NP_001230928.1 | |
NM_001244000.1 | 430 | Silent Mutation | CCA,CCG | P,P 104 | NP_001230929.1 | |
NM_003121.4 | 430 | Silent Mutation | CCA,CCG | P,P 135 | NP_003112.2 |