Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAACAATCACATCTTTCACGTCAT[A/C]GTAGACTTCATACTGGAGAGAAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF808 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF808 - zinc finger protein 808 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039886.3 | 1794 | Intron | NP_001034975.2 | |||
NM_001321424.1 | 1794 | Intron | NP_001308353.1 | |||
NM_001321425.1 | 1794 | Intron | NP_001308354.1 | |||
XM_005258909.3 | 1794 | Missense Mutation | AGT,CGT | S,R 517 | XP_005258966.1 |