Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGCTGGAAGGAGGCCCAGCCAGG[A/G]TCCAGGGTGTGGGAGGCTGTCCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614566 MIM: 191044 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAAF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAAF3 - dynein axonemal assembly factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256714.1 | Intron | NP_001243643.1 | ||||
NM_001256715.1 | Intron | NP_001243644.1 | ||||
NM_001256716.1 | Intron | NP_001243645.1 | ||||
NM_178837.4 | Intron | NP_849159.2 |
TNNI3 - troponin I3, cardiac type | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |