Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCCCAGCAGACAGGCCCTTGGG[A/G]GCATGTGGAGGGCTGAGCAAAAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609235 MIM: 612939 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRSK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRSK1 - BR serine/threonine kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032430.1 | Intron | NP_115806.1 | ||||
XM_005259327.3 | Intron | XP_005259384.1 | ||||
XM_011527395.2 | Intron | XP_011525697.2 |
HSPBP1 - HSPA (Hsp70) binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |