Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGCCAAGGGTCCTTTTACTCAGT[A/G]GCAGCAACAAACGCAGTCTGTTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603638 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6805 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR6805 - microRNA 6805 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL28 - ribosomal protein L28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000991.4 | 310 | Intron | NP_000982.2 | |||
NM_001136134.1 | 310 | Intron | NP_001129606.1 | |||
NM_001136135.1 | 310 | Intron | NP_001129607.1 | |||
NM_001136136.1 | 310 | Intron | NP_001129608.1 | |||
NM_001136137.1 | 310 | UTR 3 | NP_001129609.1 |
TMEM190 - transmembrane protein 190 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM238 - transmembrane protein 238 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |