Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGCACAATAAATCTTTCTCAGGA[C/T]CTAAGACACTTAAAAATATAATCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601483 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PEG3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PEG3 - paternally expressed 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146184.1 | 5293 | UTR 3 | NP_001139656.1 | |||
NM_001146185.1 | 5293 | UTR 3 | NP_001139657.1 | |||
NM_001146186.1 | 5293 | UTR 3 | NP_001139658.1 | |||
NM_001146187.1 | 5293 | UTR 3 | NP_001139659.1 | |||
NM_006210.2 | 5293 | UTR 3 | NP_006201.1 |
PEG3-AS1 - PEG3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZIM2 - zinc finger imprinted 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146326.1 | 5293 | Intron | NP_001139798.1 | |||
NM_001146327.1 | 5293 | Intron | NP_001139799.1 | |||
NM_015363.4 | 5293 | Intron | NP_056178.3 |