Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAATTTTTTTTAAGTGTTCTAGTT[A/G]TTGGTTAGATGTGAAGTTTCCATAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF772 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF772 - zinc finger protein 772 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024596.2 | 10893 | Intron | NP_001019767.1 | |||
NM_001144068.1 | 10893 | Intron | NP_001137540.1 | |||
XM_005258943.4 | 10893 | UTR 3 | XP_005259000.1 | |||
XM_005258944.3 | 10893 | Intron | XP_005259001.1 | |||
XM_011526980.2 | 10893 | UTR 3 | XP_011525282.1 | |||
XM_017026813.1 | 10893 | UTR 3 | XP_016882302.1 |