Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCGAGCGGGCCGCACCTCGCGCT[A/G]GGCCAGCTCTGGACGCTGTGCCGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZSCAN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZSCAN1 - zinc finger and SCAN domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182572.3 | 2028 | Silent Mutation | CTA,CTG | L,L 56 | NP_872378.3 | |
XM_006723149.2 | 2028 | Silent Mutation | CTA,CTG | L,L 96 | XP_006723212.1 | |
XM_017026609.1 | 2028 | Silent Mutation | CTA,CTG | L,L 96 | XP_016882098.1 | |
XM_017026610.1 | 2028 | Silent Mutation | CTA,CTG | L,L 56 | XP_016882099.1 |