Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAGTTTATAACCCGGGAAGAGCT[A/G]CTCTCAGAGCGGAGGGAGACCATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF324 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF324 - zinc finger protein 324 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF446 - zinc finger protein 446 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304453.1 | 51 | Intron | NP_001291382.1 | |||
NM_017908.3 | 51 | Intron | NP_060378.1 | |||
XM_005259052.4 | 51 | UTR 5 | XP_005259109.1 | |||
XM_005259053.4 | 51 | UTR 5 | XP_005259110.1 | |||
XM_006723266.3 | 51 | UTR 5 | XP_006723329.1 | |||
XM_017026949.1 | 51 | UTR 5 | XP_016882438.1 | |||
XM_017026950.1 | 51 | UTR 5 | XP_016882439.1 |