Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTTTGTGTCTTATCAAAAATCA[A/G]TTGACTATATGTGTGATTATATTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUDT19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUDT19 - nudix hydrolase 19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TDRD12 - tudor domain containing 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110822.1 | Intron | NP_001104292.1 | ||||
XM_011527471.2 | Intron | XP_011525773.1 | ||||
XM_011527472.2 | Intron | XP_011525774.1 | ||||
XM_011527473.2 | Intron | XP_011525775.1 | ||||
XM_011527474.2 | Intron | XP_011525776.1 | ||||
XM_011527475.2 | Intron | XP_011525777.1 | ||||
XM_011527476.2 | Intron | XP_011525778.1 | ||||
XM_017027458.1 | Intron | XP_016882947.1 | ||||
XM_017027459.1 | Intron | XP_016882948.1 |