Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCTATGTTCTCTACCTCCCGTG[C/T]CTTGACCAACCTCAGATTTATACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF30 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF30 - zinc finger protein 30 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099437.1 | 245 | Intron | NP_001092907.1 | |||
NM_001099438.1 | 245 | Intron | NP_001092908.1 | |||
NM_194325.2 | 245 | Intron | NP_919306.2 | |||
XM_011527443.2 | 245 | Intron | XP_011525745.1 | |||
XM_011527444.1 | 245 | Intron | XP_011525746.1 | |||
XM_017027423.1 | 245 | UTR 5 | XP_016882912.1 | |||
XM_017027424.1 | 245 | Intron | XP_016882913.1 | |||
XM_017027425.1 | 245 | Intron | XP_016882914.1 | |||
XM_017027426.1 | 245 | Intron | XP_016882915.1 | |||
XM_017027427.1 | 245 | Intron | XP_016882916.1 | |||
XM_017027428.1 | 245 | Intron | XP_016882917.1 | |||
XM_017027429.1 | 245 | Intron | XP_016882918.1 |
ZNF30-AS1 - ZNF30 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |