Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGAGTGGTGGCTACAGTGACCGG[A/G]GCTCGGGCGGGTCCTACAGAGACAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602649 | ||||||||||||||||||||
Literature Links: |
C19orf24 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C19orf24 - chromosome 19 open reading frame 24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CIRBP - cold inducible RNA binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001280.2 | 614 | Missense Mutation | AGC,GGC | S,G 155 | NP_001271.1 | |
NM_001300815.1 | 614 | Missense Mutation | AGC,GGC | S,G 155 | NP_001287744.1 | |
NM_001300829.1 | 614 | Missense Mutation | AGC,GGC | S,G 155 | NP_001287758.1 | |
XM_006722637.1 | 614 | Missense Mutation | AGC,GGC | S,G 155 | XP_006722700.1 | |
XM_011527668.1 | 614 | Missense Mutation | AGC,GGC | S,G 155 | XP_011525970.1 | |
XM_017026237.1 | 614 | Missense Mutation | AGC,GGC | S,G 155 | XP_016881726.1 |
CIRBP-AS1 - CIRBP antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |