Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTTCAGAAGCATGACCAGTGAGA[A/C]GTCCCTAGGGCCCAGGCCGGGCTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612034 MIM: 600487 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APC2 - APC2, WNT signaling pathway regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005883.2 | 1606 | Intron | NP_005874.1 | |||
XM_005259475.2 | 1606 | Intron | XP_005259532.1 | |||
XM_006722607.2 | 1606 | Intron | XP_006722670.1 | |||
XM_006722608.3 | 1606 | Intron | XP_006722671.2 | |||
XM_006722609.3 | 1606 | Intron | XP_006722672.1 | |||
XM_006722610.3 | 1606 | Intron | XP_006722673.2 |
C19orf25 - chromosome 19 open reading frame 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152482.2 | 1606 | UTR 3 | NP_689695.2 | |||
XM_005259506.3 | 1606 | UTR 3 | XP_005259563.1 | |||
XM_006722653.3 | 1606 | UTR 3 | XP_006722716.1 | |||
XM_017026374.1 | 1606 | UTR 3 | XP_016881863.1 |
PCSK4 - proprotein convertase subtilisin/kexin type 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |