Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGGTTGTTCAGATGGGAGTAAAA[A/T]TACTCATTCAGCACTTCCGTCGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608127 | ||||||||||||||||||||
Literature Links: |
PBX4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PBX4 - PBX homeobox 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025245.2 | 652 | Nonsense Mutation | TAA,TAT | *,Y 228 | NP_079521.1 | |
XM_006722911.3 | 652 | Nonsense Mutation | TAA,TAT | *,Y 142 | XP_006722974.1 | |
XM_011528320.2 | 652 | Nonsense Mutation | TAA,TAT | *,Y 245 | XP_011526622.1 | |
XM_011528322.2 | 652 | Nonsense Mutation | TAA,TAT | *,Y 142 | XP_011526624.1 | |
XM_017027326.1 | 652 | Nonsense Mutation | TAA,TAT | *,Y 211 | XP_016882815.1 | |
XM_017027327.1 | 652 | Nonsense Mutation | TAA,TAT | *,Y 197 | XP_016882816.1 | |
XM_017027328.1 | 652 | Nonsense Mutation | TAA,TAT | *,Y 245 | XP_016882817.1 | |
XM_017027329.1 | 652 | Nonsense Mutation | TAA,TAT | *,Y 63 | XP_016882818.1 | |
XM_017027330.1 | 652 | Intron | XP_016882819.1 |