Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTAGACAAGGCTCAGGAAGTGCAC[A/C]AAGAACTGGAGCATGACGGTGAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609694 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP13A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP13A1 - ATPase 13A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020410.2 | 3217 | Missense Mutation | TTG,TTT | L,F 1063 | NP_065143.2 |
GMIP - GEM interacting protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |