Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCTCTGGGTAGTTGTCACACACC[A/G]GTTAAGTCCCTTGTGACCTCCTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF708 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF708 - zinc finger protein 708 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297560.1 | 403 | Intron | NP_001284489.1 | |||
NM_001297561.1 | 403 | Intron | NP_001284490.1 | |||
NM_021269.2 | 403 | Intron | NP_067092.2 | |||
XM_017027201.1 | 403 | Intron | XP_016882690.1 | |||
XM_017027202.1 | 403 | Missense Mutation | CGG,TGG | R,W 54 | XP_016882691.1 | |
XM_017027203.1 | 403 | Missense Mutation | CGG,TGG | R,W 54 | XP_016882692.1 | |
XM_017027204.1 | 403 | Missense Mutation | CGG,TGG | R,W 130 | XP_016882693.1 |