Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCAAATATTTGGAGAATAATAGT[C/T]AGGGGAGAGCTTGAGGTCCAGGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606945 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LDLR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LDLR - low density lipoprotein receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000527.4 | Intron | NP_000518.1 | ||||
NM_001195798.1 | Intron | NP_001182727.1 | ||||
NM_001195799.1 | Intron | NP_001182728.1 | ||||
NM_001195800.1 | Intron | NP_001182729.1 | ||||
NM_001195803.1 | Intron | NP_001182732.1 | ||||
XM_011528010.1 | Intron | XP_011526312.1 |