Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAGGCACACTCATAAAAGGCCTG[A/G]ATACCAGACAGGGAGGGAGAAGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601913 MIM: 603002 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASNA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASNA1 - arsA arsenite transporter, ATP-binding, homolog 1 (bacterial) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C19orf43 - chromosome 19 open reading frame 43 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNPO2 - transportin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |