Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACCTCAGGCTTCAGGTCCAGATG[A/G]GGTTGGGTTCGGGATTATGAGCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZSWIM4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZSWIM4 - zinc finger SWIM-type containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023072.2 | Intron | NP_075560.2 | ||||
XM_005260023.3 | Intron | XP_005260080.1 | ||||
XM_017027153.1 | Intron | XP_016882642.1 | ||||
XM_017027154.1 | Intron | XP_016882643.1 | ||||
XM_017027155.1 | Intron | XP_016882644.1 | ||||
XM_017027156.1 | Intron | XP_016882645.1 | ||||
XM_017027157.1 | Intron | XP_016882646.1 |