Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGCTCACCTGCAGAAAGTCAGC[C/T]GTCTGCTTAGGGAGCTTGGTGCCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609475 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKAP8L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AKAP8L - A-kinase anchoring protein 8 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291478.1 | 1389 | Silent Mutation | ACA,ACG | T,T 367 | NP_001278407.1 | |
NM_014371.3 | 1389 | Silent Mutation | ACA,ACG | T,T 428 | NP_055186.3 | |
XM_005259854.3 | 1389 | Silent Mutation | ACA,ACG | T,T 394 | XP_005259911.2 | |
XM_011527916.2 | 1389 | Silent Mutation | ACA,ACG | T,T 428 | XP_011526218.1 |