Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCAGGGGCCACGGTGGGAGGCCC[A/G]TCCTCAGCCAGGGCAGGGGCCAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609475 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKAP8L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AKAP8L - A-kinase anchoring protein 8 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WIZ - widely interspaced zinc finger motifs | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021241.2 | 2613 | Silent Mutation | GAC,GAT | D,D 440 | NP_067064.2 | |
XM_005260004.4 | 2613 | Silent Mutation | GAC,GAT | D,D 1535 | XP_005260061.1 | |
XM_005260005.4 | 2613 | Silent Mutation | GAC,GAT | D,D 1478 | XP_005260062.1 | |
XM_005260006.4 | 2613 | Silent Mutation | GAC,GAT | D,D 1402 | XP_005260063.1 | |
XM_005260007.4 | 2613 | Silent Mutation | GAC,GAT | D,D 1345 | XP_005260064.1 | |
XM_005260008.2 | 2613 | Silent Mutation | GAC,GAT | D,D 798 | XP_005260065.1 | |
XM_005260009.2 | 2613 | Silent Mutation | GAC,GAT | D,D 614 | XP_005260066.1 | |
XM_005260010.3 | 2613 | Silent Mutation | GAC,GAT | D,D 614 | XP_005260067.1 | |
XM_005260011.2 | 2613 | Silent Mutation | GAC,GAT | D,D 608 | XP_005260068.1 | |
XM_005260012.2 | 2613 | Silent Mutation | GAC,GAT | D,D 481 | XP_005260069.1 | |
XM_006722828.3 | 2613 | Silent Mutation | GAC,GAT | D,D 761 | XP_006722891.1 | |
XM_011528163.2 | 2613 | Silent Mutation | GAC,GAT | D,D 614 | XP_011526465.1 |