Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGATGATGGGGCTGAGGAAGGGT[C/G]TGAGGACCGTGTAGGTGATGCCCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
OR10H1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OR10H1 - olfactory receptor family 10 subfamily H member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013940.2 | 937 | Missense Mutation | ACA,AGA | T,R 283 | NP_039228.1 | |
XM_011527911.1 | 937 | Missense Mutation | ACA,AGA | T,R 283 | XP_011526213.1 |