Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCACCTGTTCCAGCTCCCTGCGCA[A/G]AGACTGCAGCTGCTCCTGCAGGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 132880 MIM: 611810 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NR2F6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NR2F6 - nuclear receptor subfamily 2 group F member 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USHBP1 - USH1 protein network component harmonin binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297703.1 | 1856 | Silent Mutation | CTG,TTG | L,L 547 | NP_001284632.1 | |
NM_001321417.1 | 1856 | Silent Mutation | CTG,TTG | L,L 611 | NP_001308346.1 | |
NM_031941.3 | 1856 | Silent Mutation | CTG,TTG | L,L 611 | NP_114147.2 | |
XM_011528351.2 | 1856 | Intron | XP_011526653.1 |