Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGAATTCTAAAGGAACTGGAACA[C/T]GTGAAGGCTTTCCCACACTCCTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF426 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF426 - zinc finger protein 426 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300883.2 | 1760 | Silent Mutation | ACA,ACG | T,T 476 | NP_001287812.1 | |
NM_001318055.1 | 1760 | Intron | NP_001304984.1 | |||
NM_001318056.1 | 1760 | Intron | NP_001304985.1 | |||
NM_024106.2 | 1760 | Intron | NP_077011.1 | |||
XM_017027293.1 | 1760 | Silent Mutation | ACA,ACG | T,T 476 | XP_016882782.1 | |
XM_017027294.1 | 1760 | Silent Mutation | ACA,ACG | T,T 272 | XP_016882783.1 |