Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCTTCCTTGTGAGTCCAAGGCCA[A/G]GTCTCAGGACAGCCACAGACAGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607451 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GMEB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GMEB2 - glucocorticoid modulatory element binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012384.4 | 2638 | UTR 3 | NP_036516.1 | |||
XM_005260202.4 | 2638 | UTR 3 | XP_005260259.1 | |||
XM_006723776.2 | 2638 | UTR 3 | XP_006723839.1 | |||
XM_011528779.2 | 2638 | UTR 3 | XP_011527081.1 |