Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAGCGAGAGAATGGTTTGGCAGC[C/G]TTACCCAAATATTTGTTATTTTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607589 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFT52 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IFT52 - intraflagellar transport 52 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303458.2 | Intron | NP_001290387.1 | ||||
NM_001303459.2 | Intron | NP_001290388.1 | ||||
NM_001323578.1 | Intron | NP_001310507.1 | ||||
NM_001323579.1 | Intron | NP_001310508.1 | ||||
NM_001323580.1 | Intron | NP_001310509.1 | ||||
NM_001323581.1 | Intron | NP_001310510.1 | ||||
NM_016004.4 | Intron | NP_057088.2 | ||||
XM_017027863.1 | Intron | XP_016883352.1 |
SGK2 - SGK2, serine/threonine kinase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |