Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCCAGGCTTCCCGCTGGAGGAT[C/T]TCACCTGTGGATGGAGTTCCGAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602900 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNMT3B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNMT3B - DNA methyltransferase 3 beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207055.1 | Intron | NP_001193984.1 | ||||
NM_001207056.1 | Intron | NP_001193985.1 | ||||
NM_006892.3 | Intron | NP_008823.1 | ||||
NM_175848.1 | Intron | NP_787044.1 | ||||
NM_175849.1 | Intron | NP_787045.1 | ||||
NM_175850.2 | Intron | NP_787046.1 | ||||
XM_011528653.2 | Intron | XP_011526955.1 | ||||
XM_011528654.2 | Intron | XP_011526956.1 |