Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAAACAAAACAAAACAAAACCCC[A/C]AAAAAAGAACCAACTGAACAGGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607662 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPATA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPATA2 - spermatogenesis associated 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135773.1 | 2064 | UTR 3 | NP_001129245.1 | |||
NM_006038.3 | 2064 | UTR 3 | NP_006029.1 | |||
XM_006723894.1 | 2064 | UTR 3 | XP_006723957.1 | |||
XM_011529116.1 | 2064 | UTR 3 | XP_011527418.1 |